Skip to Content
Merck
All Photos(1)

Key Documents

1.05323

Millipore

D(-)Fructose

suitable for microbiology

Synonym(s):

D(-)Fructose, Laevulose, Levulose

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€68.10

€68.10


Estimated to ship on30 May 2025


Request more information

Select a Size

Change View
250 G
€68.10

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
41106212
EC Index Number:
200-333-3
NACRES:
NA.74

€68.10


Estimated to ship on30 May 2025


Request more information

form

solid

Quality Level

autoignition temp.

360 °C

pH

5-6 (20 °C, 100 g/L in H2O)

mp

100-104 °C (decomposition)

solubility

790 g/L (OECD Test Guideline 105)

density

1.59 g/cm3 at 20 °C (OECD Test Guideline 109)

bulk density

700‑800 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

O[C@H]([C@H](O)CO)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Analysis Note

Identity (IR-spectrum): conforms
Spec. rotation (α 20/D; 10 %; water): -93 - -91 °
Heavy metals (as Pb): ≤ 0.001 %
TLC-Test: conforms
Water: ≤ 0.5 %
Suitability for microbiology: conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service