Skip to Content
Merck
All Photos(1)

Key Documents

1.05206

Millipore

Creatinine

for biochemistry

Synonym(s):

Creatinine

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€73.60
500 G
€575.00

€73.60


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
50 G
€73.60
500 G
€575.00

About This Item

CAS Number:
Molecular Weight:
113.11
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-466-7
NACRES:
NA.21

€73.60


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

form

solid

pH

7.0-9.0 (20 °C, 50 g/L in H2O)

mp

305 °C (decomposition)

solubility

90 g/L

bulk density

484 kg/m3

storage temp.

no temp limit

InChI

1S/C4H7N3O/c1-7-2-3(8)6-4(7)5/h2H2,1H3,(H2,5,6,8)

InChI key

DDRJAANPRJIHGJ-UHFFFAOYSA-N

General description

for biochemistry

Application

Creatinine for biochemistry. CAS 60-27-5, pH 7.0 - 9.0 (50 g/l, H₂O, 20 °C).

Analysis Note

Assay (perchloric acid titration)≥ 99.0 %Identity (IR-spectrum)passes testpH-value (5 %; water)7 - 9Spec. Absorptivity A 1%/1cm (234 nm; 0.001 %; buffer pH 7.0)620 - 640Chloride (Cl)≤ 0.2 %Heavy metals (as Pb)≤ 0.001 %Creatine (HPLC)≤ 0.2 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

554.0 °F

Flash Point(C)

290 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service