Skip to Content
Merck
All Photos(1)

Key Documents

1.04439

Supelco

Mercury(II) nitrate monohydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Mercury(II) nitrate monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€93.70
250 G
€227.00

€93.70


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€93.70
250 G
€227.00

About This Item

Linear Formula:
Hg(NO3)2 · H2O
CAS Number:
Molecular Weight:
324.60 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-152-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid

€93.70


Estimated to ship on23 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (complexometric)

form

solid

potency

26 mg/kg LD50, oral (Rat)
75 mg/kg LD50, skin (Rat)

reduction residue

≤0.01%

mp

54.6 °C (anhydrous substance)

density

3.89 g/cm3 at 20.2 °C

bulk density

1200 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.002%

cation traces

Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
Na: ≤0.005%

storage temp.

2-30°C

SMILES string

[Hg+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O

InChI

1S/Hg.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

KVICROHOONHSRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (complexometric): ≥ 99.0 %
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.002 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.005 %
Na (Sodium): ≤ 0.005 %
Residue after reduction: ≤ 0.01 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service