Skip to Content
Merck
All Photos(1)

Key Documents

1.04428

Supelco

Mercury(II) iodide

red, for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Mercury(II) iodide, Mercury diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HgI2
CAS Number:
Molecular Weight:
454.40
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-873-8
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
0.06 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

0.006 hPa ( 80 °C)

product line

EMSURE®

Assay

≥99.0% dry basis (silver nitrate titration)

form

solid

potency

18 mg/kg LD50, oral (Rat)
75 mg/kg LD50, skin (Rat)

impurities

≤0.02% Substances not reducing by formic acid

pH

6-7 (20 °C, 50 g/L in H2O, slurry)

bp

354 °C/1013 hPa (sublimed)

mp

259 °C

solubility

0.06 g/L

density

6.36 g/cm3 at 25 °C

bulk density

1350 kg/m3

cation traces

Fe: ≤0.001%
Hg+: ≤0.1%
heavy metals (as Pb): ≤0.001%
soluble mercury salts (as Hg): ≤0.05%

storage temp.

2-30°C

SMILES string

[Hg+2].[I-].[I-]

InChI

1S/Hg.2HI/h;2*1H/q+2;;/p-2

InChI key

YFDLHELOZYVNJE-UHFFFAOYSA-L

Application

  • Cation-Selective and Anion-Controlled Fluorogenic Behaviors of a Benzothiazole-Attached Macrocycle That Correlate with Structural Coordination Modes.: This study explores the fluorogenic behaviors of a benzothiazole-attached macrocycle, which exhibits cation-selective and anion-controlled properties. The research highlights the structural coordination modes that influence these behaviors, providing insights into the development of chemical sensors and analytical methods utilizing Mercury(II) iodide as a key component. (Ju et al., 2016).

Analysis Note

Assay (argentometric, dried sust.): ≥ 99.0 %
Solubility in Potassium iodide: passes test
Soluble mercury salts (as Hg): ≤ 0.05 %
Foreign heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Mercury (I) (as Hg): ≤ 0.1 %
Substances not reducing by formic acid: ≤ 0.02 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service