Skip to Content
Merck
All Photos(1)

Key Documents

1.03863

Millipore

NutriSelect® plus Anaerobic Agar

acc. To BREWER

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€253.00

€253.00


Estimated to ship on16 April 2025


Request more information

Select a Size

Change View
500 G
€253.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€253.00


Estimated to ship on16 April 2025


Request more information

Quality Level

form

powder

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

suitability

nonselective for anaerobic bacteria: Clostridium spp.

General description

NutriSelect® plus Anaerobic Agar is specifically designed for the surface cultivation of Clostridia spp and other anaerobic microorganisms. The medium is supplemented with series of reducing agents like thioglycolate, sulfoxylate formaldehydeand cystine which ensures complete anaerobiosis during the incubation period. The anaerobic environment is indicated by a redox indicator, methylene blue, which turns colorless during the absence of oxygen. The microbes are cultivated using pour plate method and should be incubated using Anaerocult systems.

Application

NutriSelect® plus Anaerobic Agar is used to detect anaerobic microorganisms like Clostridia spp in test samples using pour plate method.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service