Skip to Content
Merck
All Photos(1)

Documents

1.03790

Millipore

Linden Grain Medium

suitable for sterility testing, for yeasts, for molds, for bacteria

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

medium granules (dehydrated culture medium (DCM))

manufacturer/tradename

GranuCult® plus

application(s)

food and beverages
sterility testing

storage temp.

2-8°C

suitability

bacteria
molds
yeasts

General description

Linden Grain Medium (Base) is a granulated dehydrated culture media with high nutrient content. It is used for microbiological validation of aseptic filling lines across the beverage industry because it creates optimum growth conditions for fungi and bacteria at various pH levels. The pH of the medium can be adjusted to the filling line criteria. At a pH of 4.3, fungi and bacteria which are adapted to grow under high acid conditions show visible growth within 7 days of incubation at 28°C. Bacteria that grow best at neutral to slightly alkaline pH, like Bacillus subtilis, show no growth/very limited growth at pH 4.3. Any growth within the medium (turbidity), on the surface of the medium, or the inner wall or bottom of the tubes and bottles indicates the presence of spoilage organisms. Experiments have shown that the medium could also be used after freezing and thawing, thus making this medium suitable for ice crusher installations.

Application

Linden Grain Medium is suitable for the detection of yeasts, molds, and bacteria in beverages. It is designed for media fill simulations of beverage bottling and ice crusher installations.

Features and Benefits

The granulation feature of this Linden Grain Medium significantly reduces the dust formation during handling in the aseptic area

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service