Skip to Content
Merck
All Photos(1)

Key Documents

1.07993

Millipore

GranuCult® BAT Broth

according to IFU Method No. 12, For the enrichment of Alicyclobacillus spp.

Synonym(s):

BAT medium, Alicyclobacillus Medium

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€218.00

€218.00


Estimated to ship on21 May 2025


Request more information

Select a Size

Change View
500 G
€218.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

€218.00


Estimated to ship on21 May 2025


Request more information

Agency

according to IFU Method No. 12

Quality Level

sterility

non-sterile

form

medium granules (dehydrated culture medium (DCM))

manufacturer/tradename

GranuCult® prime

application(s)

bioburden testing
food and beverages

suitability

Alicyclobacillus spp. (spp)

General description

BAT broth is suitable for the enrichment of spore-forming thermo-acidophilic spoilage bacteria (Alicyclobacillus spp.). It contains glucose as a carbon and energy source. Yeast extract is a source for vitamins, particularly the B-group. Potassium dihydrogen phosphate acts as a buffering system. The low pH value and the high incubation temperature inhibit the contaminating flora. This medium contains many trace elements, which supply the specific requirements for the growth of Alicyclobacillus spp.

Application

BAT broth is used for the enrichment of Alicyclobacillus spp. from juice and juice-related products and their ingredients, environmental samples, including process water, and other materials in the area of food production and food handling.

Features and Benefits

The formulation of this BAT broth is according to the revision of IFU Method No. 12 of 2019, and is less toxic due to the removal of boric acid and cobalt(II) chloride hexahydrate

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

Related Content

Alicyclobacillus are acidophilic, strictly aerobic, and spore-forming spoilage bacteria that can cause increased turbidity, color changes and unpleasant sensory properties in juices. Testing is performed according to IFU method No 12 (2019).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service