Skip to Content
Merck
All Photos(1)

Key Documents

1.03590

Supelco

Diphenylamine-4-sulfonic acid sodium salt

redox indicator, ACS

Synonym(s):

Diphenylamine-4-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€300.00

€300.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
€300.00

About This Item

Empirical Formula (Hill Notation):
C12H10NNaO3S
CAS Number:
Molecular Weight:
271.27
MDL number:
UNSPSC Code:
12352107
EC Index Number:
228-165-6
NACRES:
NA.21

€300.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

Diphenylamine-4-sulfonic acid sodium salt, redox indicator ACS

grade

ACS reagent

Quality Level

form

solid

loss

≤5.0% loss on drying, 105°C

pH

5.7 (20 °C, 10 g/L in H2O)

mp

200 °C

solubility

820 g/L

bulk density

160 kg/m3

storage temp.

15-25°C

SMILES string

[Na].[S](=O)(=O)(O)c1ccc(cc1)Nc2ccccc2

InChI

1S/C12H11NO3S.Na/c14-17(15,16)12-8-6-11(7-9-12)13-10-4-2-1-3-5-10;/h1-9,13H,(H,14,15,16);

InChI key

VMVSDVIYIMNZBS-UHFFFAOYSA-N

Analysis Note

Appearance of solution: conforms
Loss on Drying (105°C): ≤ 5.0 %
Suitability as redox indicator: conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service