Skip to Content
Merck
All Photos(1)

Documents

1.03098

Supelco

2,2′-Bipyridine

GR for analysis (reagent for iron(II) and molybdenum) ACS

Synonym(s):

2,2′-Bipyridine, Reagents for Iron(II) and molybdenum, α,α′-Dipyridyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2
CAS Number:
Molecular Weight:
156.18
MDL number:
UNSPSC Code:
12352005
EC Index Number:
206-674-4
NACRES:
NB.21

grade

ACS reagent

Quality Level

form

solid

potency

100 mg/kg LD50, oral (Rat)
625-1250 mg/kg LD50, skin (Rat)

pH

7.5 (25 °C, 5 g/L in H2O)

bp

272 °C/1013 hPa

mp

70-72 °C

transition temp

flash point 121 °C

bulk density

500 kg/m3

storage temp.

2-30°C

InChI

1S/C10H8N2/c1-3-7-11-9(5-1)10-6-2-4-8-12-10/h1-8H

InChI key

ROFVEXUMMXZLPA-UHFFFAOYSA-N

Application


  • A novel high-throughput analytical method to quantify microplastics in water by flow cytometry.: In this paper water is used to wash the Natural aquarium silica sand and also to prepare the model for the removal of microplastics. (Li et al., 2023).


Analysis Note

Assay (perchloric acid titration): ≥ 99.5 %
Identity (IR-spectrum): passes test
Melting point: 70 - 72 °C
Absorbance of solution (50% w/v; methanol): conforms
Sensitivity (to ferrous ion): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

267.8 °F - closed cup

Flash Point(C)

131 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service