Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
250 g | Check Cart for Availability | € 241,00 |
About This Item
Quality Segment
product line
EMSURE®
assay
≥98.0% (iodometric)
form
solid
potency
3250 mg/kg LD50, oral (Rat)
impurities
≤0.05% Substances not precipitated by ammonia (as sulfates)
pH
2-3 (20 °C, 50 g/L in H2O)
mp
36-37 °C
solubility
810 g/L
bulk density
710 kg/m3
anion traces
chloride (Cl-): ≤0.002%, sulfate (SO42-): ≤0.005%
cation traces
Cu: ≤0.001%, Fe: ≤0.01%, NH4+: ≤0.002%
storage temp.
15-25°C
SMILES string
[Cr](O[N+](=O)[O-])(O[N+](=O)[O-])O[N+](=O)[O-]
InChI
1S/Cr.3NO3/c;3*2-1(3)4/q+3;3*-1
InChI key
PHFQLYPOURZARY-UHFFFAOYSA-N
Analysis Note
pH-value (5 %; water): 2.0 - 3.0
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.01 %
NH₄ (Ammonium): ≤ 0.002 %
Substances not precipitated by ammonia (as sulfates): ≤ 0.05 %
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form solid | form crystalline | form solid |
| solubility 810 g/L | solubility - | solubility H2O: very soluble(lit.) | solubility - |
| assay ≥98.0% (iodometric) | assay 99.0-101.0% (complexometric) | assay ≥99.99% trace metals basis | assay - |
| Quality Level 300 | Quality Level 300 | Quality Level 100 | Quality Level 200 |
| anion traces chloride (Cl-): ≤0.002%, sulfate (SO42-): ≤0.005% | anion traces chloride (Cl-): ≤0.0005%, phosphate (PO43-): ≤0.005%, sulfate (SO42-): ≤0.005% | anion traces - | anion traces - |
| impurities ≤0.05% Substances not precipitated by ammonia (as sulfates) | impurities ≤0.005% Insoluble matter, ≤0.1% Substances not precipitated by ammonia (as sulfate), ≤0.3% Free acid (as HNO3) | impurities ≤100.0 ppm Trace Metal Analysis | impurities - |
Still not finding the right product?
Explore all of our products under Chromium(III) nitrate nonahydrate
signalword
Warning
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2 - Skin Sens. 1B
Storage Class
5.1B - Oxidizing hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




