Skip to Content
Merck
All Photos(1)

Key Documents

1.03883

Supelco

Iron(III) nitrate nonahydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Iron(III) nitrate nonahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€60.60
1 KG
€152.00

€60.60


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€60.60
1 KG
€152.00

About This Item

Linear Formula:
Fe(NO3)3 · 9 H2O
CAS Number:
Molecular Weight:
404.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-899-5
NACRES:
NA.21
grade:
ACS reagent
form:
solid

€60.60


Estimated to ship on25 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-101.0% (complexometric)

form

solid

potency

3250 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter
≤0.1% Substances not precipitated by ammonia (as sulfate)
≤0.3% Free acid (as HNO3)

pH

1.3 (20 °C, 100 g/L in H2O)

mp

47 °C

density

1.68 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤0.005%
K: ≤0.005%
Mg: ≤0.001%
Mn: ≤0.02%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.001%

storage temp.

2-30°C

SMILES string

[Fe+3].[N+](=O)([O-])O.[N+](=O)([O-])O.[N+](=O)([O-])O.O.O.O.O.O.O.O.O.O

InChI

1S/Fe.3HNO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;3*(H,2,3,4);9*1H2/q+3;;;;;;;;;;;;

InChI key

MUSSTCMSWRZDLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Linkage

Replaces: FX0225-4; FX0225-1; FX0225

Analysis Note

Assay (complexometric): 99.0 - 101.0 %
Insoluble matter: ≤ 0.005 %
Free acid (as HNO₃): ≤ 0.3 %
Chloride (Cl): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.005 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.001 %
Mn (Manganese): ≤ 0.02 %
Na (Sodium): ≤ 0.005 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.001 %
Substances not precipitated by ammonia (as sulfate): ≤ 0.1 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service