Skip to Content
Merck
All Photos(1)

Key Documents

1.01645

Supelco

Buffer solution

(di-sodium tetraborate), traceable to SRM from NIST and PTB pH 9.22 (20°C) Certipur®

Synonym(s):

TM=["CERTIPUR"] buffer solution, di-sodium tetraborate buffer, pH 9.22 Buffer solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€79.80

€79.80


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€79.80

About This Item

UNSPSC Code:
12161705
NACRES:
NA.25

€79.80


Estimated to ship on24 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

pH

9.2 (25 °C in H2O)

bp

100 °C/1013 hPa

density

1.00 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The ready-to-use Certipur® buffer solutions are prepared gravimetrically in our ISO/IEC 17025 accredited calibration laboratory from high-purity salts, acids and bases diluted in high-purity water. All balances are regularly calibrated with analytical weight sets traceable to primary standard reference materials from NIST and PTB. The pH value is measured to the highest precision using a combined glass electrode after 5-point calibration (according to DIN 19268) against reference buffer solutions (according to DIN 19266, IUPAC, NIST, Ph.Eur. and USP).

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Certipur® buffer solution (di-sodium tetraborate buffer; pH 9.22 at 20 °C) is intended for use as a calibration standard for pH instruments or pH electrodes or as a control sample for measuring the pH value.

Features and Benefits

  • The product is delivered with a comprehensive, batch-specific Certificate of Analysis showing the relevant data for the quality management system.
  • It is a Certified Reference Material (CRM) offered in solution format.
  • Intended for extremely precise calibration, qualification and monitoring of pH instruments and for testing other buffer solutions.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service