Skip to Content
Merck
All Photos(1)

Key Documents

1.01299

Millipore

Enterococci 100

Readycult®

Sign Into View Organizational & Contract Pricing

Select a Size

20 TESTS
€135.00

€135.00


Estimated to ship on17 May 2025

Consider alternative product 1102940500

Request more information

Select a Size

Change View
20 TESTS
€135.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

€135.00


Estimated to ship on17 May 2025

Consider alternative product 1102940500

Request more information

Quality Level

sterility

sterile; γ-irradiated

form

powder (dehydrated (DCM))

manufacturer/tradename

ReadyCult®

technique(s)

microbiological culture: suitable

pH

7.3-7.7 (25 °C, 17.8 g/L in H2O, after autoclaving)

solubility

17.8 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Enterococcus spp.

Linkage

Replaces: 1.01299.0002

Analysis Note

Appearance
- clearness:
clear
- colour: yellowish to yellowish-brown
pH-value (25 °C): 7.3 - 7.7
Inoculum on reference medium
- Enterococcus faecalis ATCC 19433 (WDCM 00009):
10 - 100
- Enterococcus durans ATCC 19432: 10 - 100
- Staphylococcus aureus ATCC 25923 (WDCM 00034): ≥ 1000
Growth
- Enterococcus faecalis ATCC 19433 (WDCM 00009):
fair to good
- Enterococcus durans ATCC 19432: fair to good
- Staphylococcus aureus ATCC 25923 (WDCM 00034): fair to good
Colour change to green-blue
- Enterococcus faecalis ATCC 19433 (WDCM 00009):
+
- Enterococcus durans ATCC 19432: +
- Staphylococcus aureus ATCC 25923 (WDCM 00034): -
Incubation: 24 - 48 hrs.; 35 °C; aerobic.

Legal Information

READYCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service