Skip to Content
Merck
All Photos(1)

Key Documents

1.00686

Supelco

Sulfanilic acid

GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Sulfanilic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO3S
CAS Number:
Molecular Weight:
173.19
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-482-5
NACRES:
NB.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

Assay

≥99.0-102.0% (acidimetric)

form

solid

autoignition temp.

>400 °C

potency

12300 mg/kg LD50, oral (Rat)

pH

2.5 (20 °C, 10 g/L in H2O)

mp

288 °C (decomposes)

solubility

water: 10 g/L at 20 °C

density

1.485 g/cm3 at 25 °C

bulk density

620 kg/m3

anion traces

chloride (Cl-): ≤0.002%
nitrite (NO2-): ≤0.5 ppm
sulfate (SO42-): ≤0.01%

cation traces

heavy metals (as Pb): ≤0.001%

storage temp.

15-25°C

SMILES string

[S](=O)(=O)(O)c1ccc(cc1)N

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Application


  • Modified Gas Chromatographic Analysis: Tricaprin solution is employed in modified gas chromatographic methods to determine monoacylglycerol and diacylglycerol contents in edible fats and oils, enhancing the accuracy and efficiency of lipid profiling in food science (Satou et al., 2017).

  • Emulsification of Various Triglycerides: Tricaprin solution is also integral in research studying the emulsification properties of various triglycerides using dialkylphosphatidylcholine and egg yolk lecithin, which is pivotal for understanding and improving food emulsifiers and pharmaceutical excipients (Nii and Ishii, 2005).

  • Enzyme Properties in Biotechnology: Additionally, Tricaprin is utilized in the study of lipase properties from Pseudomonas fragi, contributing to biotechnological applications in enzyme engineering and industrial enzyme applications (Lu and Liska, 1969).



Analysis Note

Assay (acidimetric): 99.0 - 102.0 %
Identity (IR-spectrum): passes test
Matter insoluble in sodium carbonate solution: ≤ 0.01 %
Chloride (Cl): ≤ 0.002 %
Nitrite (NO₂): ≤ 0.5 ppm
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
Sulfated ash: ≤ 0.01 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service