Skip to Content
Merck
All Photos(1)

Documents

1.00382

Millipore

DL-Malic acid

for biochemistry

Synonym(s):

DL-Malic acid, DL-Hydroxysuccinic acid, Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O5
CAS Number:
Molecular Weight:
134.09
MDL number:
UNSPSC Code:
12352106
EC Index Number:
230-022-8
NACRES:
NA.21

Quality Level

Assay

≥99.5% (acidimetric)

form

solid

autoignition temp.

349 °C

potency

>3200 mg/kg LD50, oral (Rat)

pH

2.3 (20 °C, 10 g/L in H2O)

mp

128-132 °C

transition temp

flash point 203 °C

density

1.60 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

2-30°C

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)

InChI key

BJEPYKJPYRNKOW-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.5 %
Identity (IR-spectrum): passes test
Melting point: 128 - 132 °C
Optical rotation (20 %; water): -0.1 - 0.1 °
Fe (Iron): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Related substances (HPLC) (Impurity A (fumaric acid)): ≤ 1.0 %
Related substances (HPLC) (Impurity B (maleic acid)): ≤ 0.05 %
Related substances (HPLC) (any other impurity): ≤ 0.1 %
Related substances (HPLC) (Sum of all other impurities): ≤ 0.5 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

397.4 °F

Flash Point(C)

203 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service