Skip to Content
Merck
All Photos(4)

Key Documents

M0875

Sigma-Aldrich

DL-Malic acid

≥98% (capillary GC)

Synonym(s):

(±)-2-Hydroxysuccinic acid, DL-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€45.00
500 G
€56.30
1 KG
€82.10

€45.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€45.00
500 G
€56.30
1 KG
€82.10

About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723539
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€45.00


Check Cart for Availability

Request a Bulk Order

vapor density

4.6 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥98% (capillary GC)

autoignition temp.

644 °F

mp

131-133 °C (lit.)

solubility

H2O: 50 mg/mL, clear, colorless to faintly yellow

SMILES string

OC(CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)

InChI key

BJEPYKJPYRNKOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

DL-Malic acid can be used as a reactant to synthesize:
  • Hydroxy esters via esterification with alcohols[1].
  • Biodegradable polymers via thermal condensation with citric acid[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

397.4 °F

Flash Point(C)

203 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorus ylide based functionalizations of tetronic and tetramic acids
Schobert R, et al.
Synthesis, 2006, 3902-3914 (2006)
Development of green/biodegradable polymers for water scaling applications
Popuri S, et al.
International Biodeterioration & Biodegradation, 95, 225-231 (2014)
G Anneroth et al.
Scandinavian journal of dental research, 88(3), 214-218 (1980-06-01)
Two saliva stimulating drugs, solutions of Hybrin and malic acid were in an in vitro study found to have a heavy demineralizing effect on the root surface of the dental hard tissue. Consequently the drugs cannot be recommended to patients
Frédéric Bigey et al.
Current biology : CB, 31(4), 722-732 (2020-12-11)
Production of leavened bread dates to the second millennium BCE. Since then, the art of bread making has developed, yet the evolution of bread-associated microbial species remains largely unknown. Nowadays, leavened bread is made either by using a pure commercial
Hugo Oliveira et al.
PloS one, 9(10), e108376-e108376 (2014-10-08)
Resistance rates are increasing among several problematic Gram-negative pathogens, a fact that has encouraged the development of new antimicrobial agents. This paper characterizes a Salmonella phage endolysin (Lys68) and demonstrates its potential antimicrobial effectiveness when combined with organic acids towards

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service