Skip to Content
Merck
All Photos(2)

Key Documents

100073

Sigma-Aldrich

15-PGDH Inhibitor

The 15-PGDH Inhibitor controls the biological activity of 15-PGDH. This small molecule/inhibitor is primarily used for Membrane applications.

Synonym(s):

15-PGDH Inhibitor, 5-(4-(2-Cyclohexylethoxy)-3-methoxybenzylidene)thiazolidine-2,4-dione

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€150.00

€150.00


Estimated to ship on16 April 2025



Select a Size

Change View
10 MG
€150.00

About This Item

Empirical Formula (Hill Notation):
C19H23NO4S
CAS Number:
Molecular Weight:
361.46
UNSPSC Code:
51111800
NACRES:
NA.77

€150.00


Estimated to ship on16 April 2025


Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL, clear, pale yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC/1=O)SC1=C/C2=CC=C(OCCC3CCCCC3)C(OC)=C2

InChI

1S/C19H23NO4S/c1-23-16-11-14(12-17-18(21)20-19(22)25-17)7-8-15(16)24-10-9-13-5-3-2-4-6-13/h7-8,11-13H,2-6,9-10H2,1H3,(H,20,21,22)/b17-12-

InChI key

REFPRDWFDSQKAG-ATVHPVEESA-N

General description

A cell-permeable, thiazolidinedione derivative that acts as a 15-hydroxyprostaglandin dehydrogenase (15-PGDH) inhibitor (IC50 = 25 nM). It is shown to increase PGE2 levels in IL-1β-stimulated A549 cells, dose-dependently, in the range of 0–20 µM, and elicits cochlear blood flow (CBF) increase by 15% and 55% when applied intravenously to 250 g guinea pigs at 10 µg and 20 µg respectively.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wu, Y., et al. 2011. J. Med. Chem.54, 5260.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service