Skip to Content
Merck
All Photos(1)

Key Documents

09-819

Sigma-Aldrich

Anti-Lin28 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein lin-28 homolog A, Lin-28A, Zinc finger CCHC domain-containing protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€296.00

€296.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€296.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€296.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LIN28A(79727)

General description

Lin28 is a developmentally regulated RNA binding protein (RBP). Its down regulation is mediated in part by Lin4 and miR-125b, the mammalian micro-RNA homolog of Lin4. It is known to be negatively regulated by retinoic acid and act as a translational enhancer by driving specific mRNAs to polysomes and thus increasing the efficiency of protein synthesis by increasing the number of initiation events per mRNA molecule. Lin28 is necessary and sufficient for selectively blocking the cleavage processing of pri-let-7 miRNAs in embryonic cells. Lin28 is a negative regulator of miRNA biogenesis and may play a role in blocking miRNA-mediated differentiation in stem cells and in certain cancer. The mechanism by which miR-125b represses Lin28 in mammalian cells appears to involve reductions not only in translational efficiency, but also in mRNA abundance. Lin28 has shown to be essential for skeletal muscle differentiation through the translational upregulation of IGF2 expression. It is known to be expressed in testis, placenta, and embryonic stem (ES) cells.

Specificity

Other homologies: Rat and Mouse 98% and 97% sequence homology

Immunogen

Histidine-tagged recombinant protein corresponding to human Lin28.

Application

Anti-Lin28 Antibody is a Rabbit Polyclonal Antibody for detection of Lin28 also known as Protein lin-28 homolog A & has been validated in WB.

Quality

Evaluated by Western Blot in NT2/D1 cell lyate.

Western Blot Analysis: 1:5,000 dilution of this antibody detected Lin28 in 10 µg of NT2/D1 cell lysate.

Target description

~28 kDa observed

Analysis Note

Control
NT2/D1 cell lyate

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service