Skip to Content
Merck
All Photos(1)

Key Documents

07-1385

Sigma-Aldrich

Anti-Lin28 Antibody

serum, from rabbit

Synonym(s):

RNA-binding protein LIN-28

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€421.00

€421.00


Estimated to ship on13 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on13 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LIN28A(79727)

General description

Lin28 is a developmentally regulated RNA binding protein (RBP). Its down regulation is mediated in part by Lin4 and miR-125b, the mammalian micro-RNA homolog of Lin4. It is known to be negatively regulated by retinoic acid and act as a translational enhancer by driving specific mRNAs to polysomes and thus increasing the efficiency of protein synthesis by increasing the number of initiation events per mRNA molecule. Lin28 is necessary and sufficient for selectively blocking the cleavage processing of pri-let-7 miRNAs in embryonic cells (Viswanathan, 2008). Lin28 is a negative regulator of miRNA biogenesis and may play a role in blocking miRNA-mediated differentiation in stem cells and in certain cancers (Viswanathan, 2008). The mechanism by which miR-125b represses Lin28 in mammalian cells appears to involve reductions not only in translational efficiency, but also in mRNA abundance (Wu, 2005). Lin28 has shown to be essential for skeletal muscle differentiation through the translational upregulation of IGF2 expression. It is known to be expressed in testis, placenta, and embryonic stem (ES) cells.

Specificity

Detects Lin28
Other species have not been tested.

Immunogen

Full-length His-tagged Lin28 recombinant protein (human).

Application

Anti-Lin28 Antibody detects level of Lin28 & has been published & validated for use in WB & IP.
Immunoprecipitation:
Independent lab validated this antibody in immunoprecipitation. The antibody immunoprecipitated Lin28 from Tera-2 embryonic carcinoma and mouse ES cells using 2μL of antibody per reaction.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Signaling Neuroscience

RNA Metabolism & Binding Proteins

RNA Binding Protein (RBP)

Quality

Western Blot Analysis:
This lot detected Lin28 at 1:1,000 dilution in Tera-2 embryonic carcinoma cell lysate resolved via SDS-PAGE and transferred to PVDF.

Target description

25 kDa

Physical form

Rabbit Serum
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Store at -80°C for longer term storage. Avoid repeated freeze/thaws.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Sun Lee et al.
The Journal of biological chemistry, 280(17), 16635-16641 (2005-02-22)
Micro-RNAs are small non-coding RNAs that regulate target gene expression post-transcriptionally through base pairing with the target messenger RNA. Functional characterization of micro-RNAs awaits robust experimental methods to knock-down a micro-RNA as well as to assay its function in vivo.
Geneviève Di Tomasso et al.
RNA (New York, N.Y.), 22(11), 1760-1770 (2016-09-24)
As part of their normal life cycle, most RNA molecules associate with several proteins that direct their fate and regulate their function. Here, we describe a novel method for identifying proteins that associate with a target RNA. The procedure is
Srinivas R Viswanathan et al.
Science (New York, N.Y.), 320(5872), 97-100 (2008-02-23)
MicroRNAs (miRNAs) play critical roles in development, and dysregulation of miRNA expression has been observed in human malignancies. Recent evidence suggests that the processing of several primary miRNA transcripts (pri-miRNAs) is blocked posttranscriptionally in embryonic stem cells, embryonal carcinoma cells
Ligang Wu et al.
Molecular and cellular biology, 25(21), 9198-9208 (2005-10-18)
Vertebrate genomes each encode hundreds of micro-RNAs (miRNAs), yet for few of these miRNAs is there empirical evidence as to which mRNA(s) they regulate. Here we report the identification of human lin-28 mRNA as a regulatory target of human miR-125b

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service