Skip to Content
Merck
All Photos(3)

Key Documents

07-937

Sigma-Aldrich

Anti-HDAC7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

histone deacetylase 7, histone deacetylase 7A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HDAC7(51564)

General description

Histone deacetylases (HDAC) are enzymes that regulates transcription by selectively deacetylating the eta-amino groups of lysines located near the amino termini of core histone proteins. Eight members of the HDAC family have been identified, divided in to two classes. Class I includes HDAC-1, 2, 3 and 8 while Class II includes HDAC-4, 5, 6, and 7. Human HDAC7 is composed of 912 amino acid residues. Although HDAC7 is localized mostly to the cell nucleus, it is also found in the cytoplasm, suggesting nucleo-cytoplasmic shuttling. The histone deacetylase activity of HDAC7 maps to a C-terminal domain and is dependent on interaction with class I HDACs in the nucleus. It is an active component of different transcriptional co-repressor complexes that can be recruited to specific promoter regions via interactions with transcription factors. HDAC7 catalyzes removal of acetyl-groups from acetyl-lysines of histones, resulting in a tighter DNA-histone interaction. This compaction of chromatin leads to the repression of transcription of DNA in that region.

Specificity

Not tested with other species.
This antibody recognizes human HDAC7. Does not cross-react with HDAC1, HDAC4, or HDAC8.

Immunogen

Epitope: a.a.164-182
Synthetic peptide corresponding to amino acids 164-182 of human HDAC7 [(C)KPKKSLERRKNPLLRKESA].

Application

Research Category
Epigenetics & Nuclear Function

Apoptosis & Cancer
Research Sub Category
Histones
Use Anti-HDAC7 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect HDAC7 also known as histone deacetylase 7, histone deacetylase 7A.
Western Blotting:
A previous lot of this antibody was shown to recognize recombinant HDAC7 but not recombinant HDAC1, HDAC4, or HDAC8.

Western Blotting:
A previous lot of this antibody was shown to recognize endogenous HDAC7 in HEK293T cells.

Quality

Routinely evaluated by Western blotting using recombinant HDAC7 protein.

Target description

100 kDa

Physical form

Affinity purified
Purified Rabbit polyclonal in buffer containing 20 mM PBS and 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Recombinant HDAC7 protein (Cat. #14-832).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franck Dequiedt et al.
Immunity, 18(5), 687-698 (2003-05-20)
We report that HDAC7, a class II histone deacetylase, is highly expressed in CD4(+)CD8(+) double-positive thymocytes. HDAC7 inhibits the expression of Nur77, an orphan receptor involved in apoptosis and negative selection, via the transcription factor MEF2D. HDAC7 is exported from
Histone deacetylase inhibitors selectively suppress expression of HDAC7.
Dokmanovic, Milos, et al.
Molecular Cancer Therapeutics, 6, 2525-2534 (2007)
Chengzhuo Gao et al.
Molecular and cellular biology, 28(18), 5658-5667 (2008-07-16)
Promyelocytic leukemia protein (PML) sumoylation has been proposed to control the formation of PML nuclear bodies (NBs) and is crucial for PML-dependent cellular processes, including apoptosis and transcriptional regulation. However, the regulatory mechanisms of PML sumoylation and its specific roles
Yu-yi Lin et al.
Nature, 482(7384), 251-255 (2012-02-10)
First identified as histone-modifying proteins, lysine acetyltransferases (KATs) and deacetylases (KDACs) antagonize each other through modification of the side chains of lysine residues in histone proteins. Acetylation of many non-histone proteins involved in chromatin, metabolism or cytoskeleton regulation were further

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service