Skip to Content
Merck
All Photos(1)

Key Documents

SML1719

Sigma-Aldrich

BPC 157 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

BPC-157, Booly Protection Compound 15, GEPPPGKPADDAGLV, Gly-Glu-Pro-Pro-Pro-Gly-Lys-Pro-Ala-Asp-Asp-Ala-Gly-Leu-Val

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€193.00

€193.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€193.00

About This Item

Empirical Formula (Hill Notation):
C62H98N16O22 · xC2HF3O2
CAS Number:
Molecular Weight:
1419.54 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€193.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

film

color

colorless

shipped in

wet ice

storage temp.

−20°C

SMILES string

N2([C@@H](CCC2)C(=O)N3[C@@H](CCC3)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N4[C@@H](CCC4)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(=O)O)C(=O)N[C@@H](CC(=O)O)C(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)C)C(=O)O)C(=O)[C@H]1N(CCC1)C(=O)[C@@H](NC(=O)CN)CCC(=O)O

InChI key

HEEWEZGQMLZMFE-RKGINYAYSA-N

Biochem/physiol Actions

BPC 157 is a gut peptide and possesses free radical scavenging activity. This peptide stimulates nitric oxide synthase generation and therefore offers gastric cytoprotection.[1] Unlike other peptides, BPC 157 is effective without a carrier. It participates in muscle healing and is used in the treatment of gastrocnemius muscle complex.[2]
BPC 157 is a stable gastric pentadecapeptide that exhibit anti-ulcer and wounds/fistulas healing properties.
BPC 157 is a stable gastric pentadecapeptide that exhibit anti-ulcer and wounds/fistulas healing properties. BPC 157 reduces immediate and delayed damage induced brain trauma and improves nerve regeneration after transection. BPC 157 displays antianxiety and antidepressant effects. It appears that BPC 157 promotes proliferation, migration, and tube formation of human umbilical vein endothelial cells through activation of ERK1/2 phosphorylation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gastric pentadecapeptide BPC 157 as an effective therapy for muscle crush injury in the rat.
Novinscak T, et al.
Surgery Today, 38(8), 716-725 (2008)
The influence of a novel pentadecapeptide, BPC 157, on NG-nitro-L-arginine methylester and L-arginine effects on stomach mucosa integrity and blood pressure.
Sikiric P, et al.
European Journal of Pharmacology, 332(1), 23-33 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service