Skip to Content
Merck
All Photos(1)

Key Documents

07-2054

Sigma-Aldrich

Anti-MBD1 Antibody, CT

from rabbit

Synonym(s):

CXXC-type zinc finger protein 3, Methyl-CpG-binding protein MBD1, Protein containing methyl-CpG-binding domain 1, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, the regulator of fibroblast growth factor 2 (FGF-2) <

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€360.00

€360.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€360.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

€360.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PCM1(5108)

General description

DNA methylation, or the addition of methyl groups to cytosine bases in the dinucleotide CpG, regulates gene expression and is critical for proper development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MECP2, MBD1 and MBD2 can also repress transcription from methylated gene promoters. MBD1 has been shown to play a role in chromatin modification through interaction with the histone H3K9 methyltransferase SETDB1. Five transcript variants of the MBD1 are generated by alternative splicing resulting in protein isoforms that each contain one MBD domain, two to three cysteine-rich (CXXC) domains, and some differences in the COOH terminus. MBD1, MBD2, MBD3, and MBD4 are expressed in somatic tissues, but the expression of MBD1 and MBD2 is reduced or absent in embryonic stem cells, which are known to be deficient in MeCP1 activity.

Specificity

Not tested in other species.
This antibody recognizes human MBD1.

Immunogen

Epitope: C-terminus
The immunogen was a KLH-conjugated synthetic peptide corresponding the C-terminal region of human MBD1.

Application

Anti-MBD1 Antibody, C-terminus is a Rabbit Polyclonal Antibody for detection of MBD1 also known as CXXC-type zinc finger protein 3 & has been validated in WB, ELISA.
ELISA:
This antibody has been shown by an outside laboratory to be suitable for ELISA (1:1000).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Western Blot Analysis:
This antibody recognizes MBD1 on 10 µg of HeLa cell lysate.

Target description

~61 kDa

Physical form

Format: Purified
Protein G Chromatography
Purified rabbit polyclonal in buffer containing 0.09% NaN3.

Storage and Stability

Stable for 6 months at 2-8°C from date of receipt. For long term storage, store at -20ºC. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service