Skip to Content
Merck
All Photos(2)

Documents

565794

Sigma-Aldrich

β-Secretase Inhibitor IV

InSolution, ≥95%

Synonym(s):

InSolution β-Secretase Inhibitor IV, N-(1 S, 2 R)-1-Benzyl-3-(cyclopropylamino)-2-hydroxypropyl)-5-(methyl(methylsulfonyl)amino-Nʹ-((1 R)-1-phenylethyl)isophthalamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H38N4O5S
CAS Number:
Molecular Weight:
578.72
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

dry ice

storage temp.

−70°C

SMILES string

O=C(C1=CC(C(N[C@@H](CC2=CC=CC=C2)[C@H](O)CNC3CC3)=O)=CC(N(S(C)(=O)=O)C)=C1)N[C@H](C)C4=CC=CC=C4

InChI

1S/C31H38N4O5S/c1-21(23-12-8-5-9-13-23)33-30(37)24-17-25(19-27(18-24)35(2)41(3,39)40)31(38)34-28(16-22-10-6-4-7-11-22)29(36)20-32-26-14-15-26/h4-13,17-19,21,26,28-29,32,36H,14-16,20H2,1-3H3,(H,33,37)(H,34,38)/t21-,28+,29-/m1/s1

InChI key

VPNIQGRFZCTBEZ-SPTGULJVSA-N

General description

A cell-permeable isophthalamide compound containing hydroxyethylamine motif that binds to BACE-1 active site and potently blocks its proteolytic activity (IC50 = 15 nM for BACE-1, human and 29 nM for sAPP_NF in HEK293-APPNFEV cells). Displays greater selectivity over other aspartyl proteases (IC50 = 0.23 µM, 7.6 µM and >50 µM for BACE-2, cathepsin D, and renin, respectively). The solid form of this compound (Cat. No. 565788) is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
β-Secretase
Reversible: yes
Target IC50: 15 nM for BACE-1, human and 29 nM for sAPP_NF in HEK293-APPNFEV cells

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (500 µg/86.4 µL) solution of β-Secretase Inhibitor IV (Cat. No. 565788) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C. For short term storage, aliquots are stable for up to 3 months at -20°C.

Other Notes

Stachel, S.J., et al. 2004. J. Med. Chem.47, 6447.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Scholz et al.
Journal of neurochemistry, 147(2), 256-274 (2018-05-29)
The initial step in the amyloidogenic cascade of amyloid precursor protein (APP) processing is catalyzed by beta-site APP-cleaving enzyme (BACE), and this protease has increased activities in affected areas of Alzheimer's disease brains. We hypothesized that altered APP processing, because

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service