Skip to Content
Merck
All Photos(1)

Key Documents

07-1601

Sigma-Aldrich

Anti-IRAK1 Antibody, CT

from rabbit

Synonym(s):

interleukin-1 receptor-associated kinase 1, Pelle homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€335.00

€335.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€335.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€335.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IRAK1(3654)

General description

IRAK1, also known as Interleukin-1 receptor associated kinase 1, belongs to the tyrosine kinase related (TKL) family and is involved with mRNA stabilization and activation of NF-kappa B. IRAK1 is located in all tissue as 2 isoforms, with 1 binding quickly and then degrading while isoform 2 is activated by kinase, resulting in a slower more sustained response to cytokines. Phosphorylation of IRAK1 leads to recruitment of the adapter protein PELI1. IRAK1 utilizes 2 pathways, the first one allows for mediation of IL-1 inflammatory responses when TRAF6 is bound, which activates NF-kappa B, thereby inhibiting I kappa B alpha. A second pathway does not involve TRAF6 and results in the stabilization of mRNA.

Specificity

This antibody recognizes the C-terminal region of IRAK1.

Immunogen

Epitope: C-terminus
KLH-conjugated synthetic linear peptide corresponding to the C-terminal region of IRAK1.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling
This Anti-IRAK1 Antibody, C-terminus is validated for use in WB for the detection of IRAK1.

Quality

Evaluated by western blot in jurkat lysate.

Western Blot Analysis: A 1:50-1:500 dilution of this antibody detected IRAK1 on jurkat lysate.

Target description

~77 kDa

Physical form

Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing PBS and 0.09% sodium azide.

Storage and Stability

Stable for 6 months at 2-8ºC from date of receipt. For long term storage, store at -20ºC.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Jurkat lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service