Skip to Content
Merck
All Photos(1)

Documents

06-570-AF647

Sigma-Aldrich

Anti-phospho Histone H3 (Ser10), Alexa Fluor 647 Conjugate Antibody

0.5 mg/mL, from rabbit

Synonym(s):

H3S10P, Histone H3 (phospho Ser10), H3 histone, family 3A, H3.3A, H3 histone, family 3B, H3.3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

concentration

0.5 mg/mL

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer10)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Histone proteins are highly post-translationally modified, and Histone H3 is the most extensively modified of the five histones. Histone H3 sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes. Di- and tri-methylation of histone H3 are gene silencing marks, while removal of these marks by the demethylase genes Jmjd1a and Jmjd2c leads to gene activation. This product is a conjugated version of Cat. No. 06-570.

Immunogen

Linear peptide corresponding to Histone H3 phosphorylated at Ser10.

Application

Anti-phospho-H3 (Ser10), Alexa Fluor 647 Conjugate is a purified rabbit polyclonal antibody to detect Histone H3 phosphorylated at serine 10. This Ab is labeled with Alexa Fluor 647 and validated for use in IC. The unconjugated version of this antibody is published and proven in ICC, IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected phospho Histone H3 (Ser10) in HeLa cells.

Alexa Fluor is a registered trademark of Life Technologies.

Target description

~17 kDa observed (Refer to Cat. No. 06-570)

Physical form

Protein A purified
Purified rabbit conjugated antibody in buffer containing PBS and 15 mg/ml BSA with 0.1% Sodium Azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsu-Min Sung et al.
The Journal of cell biology, 222(12) (2023-11-13)
Current models posit that nuclear speckles (NSs) serve as reservoirs of splicing factors and facilitate posttranscriptional mRNA processing. Here, we discovered that ribotoxic stress induces a profound reorganization of NSs with enhanced recruitment of factors required for splice-site recognition, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service