Skip to Content
Merck
All Photos(1)

Key Documents

07-1594

Sigma-Aldrich

Anti-Phospho-Dnmt1(Ser714) Antibody

from rabbit

Synonym(s):

CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA MTase HsaI, DNA methyltransferase 1, DNA methyltransferase HsaI

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€462.00

€462.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
400 μL
€462.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€462.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ELISA: suitable
dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer714)

Gene Information

human ... DNMT1(1786)

General description

Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes may be linked with development of cancer. Three families of mammalian DNA methyltransferase genes have been identified which include Dnmt1, Dnmt2 and Dnmt3. Dnmt1 is constitutively expressed in proliferating cells and inactivation of Dnmt1 causes global demethylation of genomic DNA and embryonic lethality. Dnmt1 co-purifies with the retinoblastoma (Rb) tumour suppressor gene product, E2F1, and HDAC1. Dnmt1 also cooperates with Rb to repress transcription from promoters containing E2F binding sites suggesting a link between DNA methylation, histone deacetylase and sequence-specific DNA binding activity, as well as a growth-regulatory pathway that is disrupted in nearly all cancer cells. Phosphorylation of Ser714 is induced by EGF, but the role of this modification is unknown.

Specificity

Not tested in other species.
This antibody recognizes human Dnmt1 phosphorylated on Ser714.

Immunogen

Epitope: Ser714
The immunogen was a KLH-conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Ser714 of human Dnmt1.

Application

Anti-Phospho-Dnmt1(Ser714) Antibody is a rabbit polyclonal antibody for detection of Phospho-Dnmt1(Ser714) also known as CXXC finger protein 9, DNA (cytosine-5-)-methyltransferase 1 & DNA MTase HsaI has been validated in WB, ELISA, DB.
ELISA:
This antibody has been shown by an outside laboratory to be suitable for ELISA (1:1000).

Dot blotting:
This antibody has been shown by an outside laboratory to be specific for the phospho-Ser714 immunogen peptide, but not the unphosphorylated peptide, by dot blotting (1:500).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Western Blot Analysis:
1:250 dilution of this antibody detected Dnmt1 on 10 µg HeLa nuclear extract

Target description

~184 kDa

Physical form

Format: Purified
Purified
Purified rabbit polyclonal in buffer containing 0.09% NaN3.

Storage and Stability

Stable for 6 months at 2-8°C from date of receipt. For long term storage, store at -20ºC. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa nuclear extract or EGF-treated whole HeLa cell extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Cuadrado et al.
The EMBO journal, 29(12), 2014-2025 (2010-05-18)
The chromatin-remodelling complex SNF2-related CBP activator protein (SRCAP) regulates chromatin structure in yeast by modulating the exchange of histone H2A for the H2A.Z variant. Here, we have investigated the contribution of H2A.Z-mediated chromatin remodelling to mammalian cell differentiation reprogramming. We
Otto Kauko et al.
The Journal of biological chemistry, 295(13), 4194-4211 (2020-02-20)
Protein phosphatase 2A (PP2A) critically regulates cell signaling and is a human tumor suppressor. PP2A complexes are modulated by proteins such as cancerous inhibitor of protein phosphatase 2A (CIP2A), protein phosphatase methylesterase 1 (PME-1), and SET nuclear proto-oncogene (SET) that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service