Skip to Content
Merck
All Photos(1)

Key Documents

07-832

Sigma-Aldrich

Anti-phospho-FAK (Tyr861) Antibody

Upstate®, from rabbit

Synonym(s):

Focal Adhesion Kinase, pp125FAK, FADK1, PTK2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€536.00

€536.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€536.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€536.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

chicken, human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr861)

Gene Information

human ... PTK2(5747)

Specificity

Predicted to cross-react with rat based on sequence homology (100%), but have not been tested.
Recognizes phosphorylated FAK (Tyr861)

Immunogen

Peptide corresponding to amino acid region encompassing the human, mouse, rat, chicken, and frog phospho-FAK (Tyr861)

Application

Detect phospho-FAK (Tyr861) using this Anti-phospho-FAK (Tyr861) Antibody validated for use in WB & IC.

Target description

125 kDa

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Focal adhesion kinase signaling controls cyclic tensile strain enhanced collagen I-induced osteogenic differentiation of human mesenchymal stem cells.
Donald F Ward,William A Williams,Nicole E Schapiro,Genevieve L Weber,Samuel R Christy et al.
Molecular & Cellular Biomechanics : Mcb null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service