Skip to Content
Merck
All Photos(1)

Key Documents

07-1548

Sigma-Aldrich

Anti-HDAC11 Antibody

from rabbit

Synonym(s):

histone deacetylase 11

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€475.00

€475.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
400 μL
€475.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€475.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HDAC11(79885)
mouse ... Hdac11(232232)

General description

HDAC11 is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation is a marker for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. The predominantly nuclear HDAC11, which interacts with HDAC6, is weakly expressed in most tissues, and strongly expressed in brain, heart, skeletal muscle, kidney and testis. Its activity is inhibited by trapoxin, a known histone deacetylase inhibitor.

Specificity

Not tested in other species.
This antibody recognizes HDAC11.

Immunogen

Epitope: N-terminus
The immunogen was a KLH-conjugated synthetic peptide corresponding the N-terminal region of human HDAC11.

Application

ELISA:
This antibody has been shown by an outside laboratory to be suitable for ELISA (1:1000).

Immunohistochemistry:
This antibody has been shown by an outside laboratory to be suitable for histochemistry using formalin fixed, paraffin embedded human hepatocarcinoma tissue (1:50-1:100).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-HDAC11 Antibody (Rabbit Polyclonal Antibody) validated in WB, ELISA, IHC to detect HDAC11 also known as histone deacetylase 11.

Quality

Western Blot Analysis:
Detection of HDAC11 in 10 µg of Mouse Brain lysate by Western blot analysis.

Target description

~40 kDa

Physical form

Format: Purified
Protein G Chromatography
Purified rabbit polyclonal in buffer containing 0.09% NaN3.

Storage and Stability

Stable for 6 months at 2-8°C from date of receipt. For long term storage, store at -20ºC. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse Brain cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service