Skip to Content
Merck
All Photos(2)

Key Documents

07-1485

Sigma-Aldrich

Anti-JAK1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Janus Kinase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€403.00

€403.00


Estimated to ship on06 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€403.00


Estimated to ship on06 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... JAK1(3716)

General description

JAKs are non-receptor tyrosine kinases which couple to cytokine receptors, and are activated by cytokines. They regulate a spectrum of cellular functions downstream of activated cytokine receptors in the lympho-hematopoietic system. JAK kinases (JAK1, JAK2, JAK3, Tyk2) have a kinase domain and a second, pseudokinase domain which is not enzymatically active. The principal effectors of JAKs are the STATs (Signal Transducers and Activators of Transcription), which are direct activators of transcription, but JAKs also signal to the Ras-MAP Kinase pathway, and through Tec, to the PI3 Kinase pathway. Immunological stimuli, such as interferons and cytokines, induce recruitment of STAT transcription factors to cytokine receptor-associated JAK2. JAK2 then phosphorylates proximal STAT transcription factors, which subsequently dimerize and translocate to the nucleus where they bind DNA. The canonical JAK/Stat pathway is integral to maintaining a normal immune system by stimulating proliferation, differentiation, survival and host resistance to pathogens. Altering JAK/Stat signaling to reduce cytokine induced pro-inflammatory responses represents an attractive target for anti-inflammatory therapies. Within the JAK2 kinase domain, there is a region that has considerable sequence homology to the regulatory region of the insulin receptor. Among a variety of sites, Tyrosines 1007 and 1008 (Tyr1007/1008) are sites of trans- or autophosphorylation in vivo and in vitro kinase reactions. JAK2 is required for signaling through a different subset of cytokine receptors, while JAK1 is essential for signaling from the IL-2, IL-6 and interferon receptors. JAK3 is required for signaling through cytokine receptors which share a common -chain.

Immunogen

Synthetic peptide corresponding to amino acids 1123-1141 of human JAK1 (CQPSNRTSFQNLIEGFEALLK)

Application

Detect JAK1 using this Anti-JAK1 Antibody validated for use in WB.

Quality

Routinely evaluated by Western Blot on PC12 lysates.

Western Blot Analysis: 1:500 dilution of this lot detected JAK1 on 10 μg of PC12 lysates.

Target description

130 kDa

Linkage

Replaces: AB3894

Physical form

Purified rabbit polyclonal IgG in 0.02 M Phosphate with 0.15 M NaCl, pH 7.6 and 0.1% sodium azide as a preservative.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service