Skip to Content
Merck
All Photos(1)

Key Documents

04-973

Sigma-Aldrich

Anti-CD39 Antibody, clone 5F2

clone 5F2, from mouse

Synonym(s):

ectonucleoside triphosphate diphosphohydrolase 1, ATPDase, ecto-apyrase, NTPDase-1, Ecto-ATP diphosphohydrolase, Lymphoid cell activation antigen, NTPDase 1, Ecto-apyrase, CD39 antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€286.00

€286.00


Estimated to ship on22 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€286.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€286.00


Estimated to ship on22 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5F2, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Entpd1(12495)

General description

Ectonucleoside triphosphate diphosphohydrolase 1 (NTDase 1 or CD39) belongs to the membrane-bound ecto-nucleoside triphosphates diphosphohydrolase family (E-NTPDases) that are characterized for their ability to hydrolyze nucleoside di- and triphosphates, such as ATP, in the presence of cations, i.e. CA2+ and Mg2+. CD39 is an important membrane protein containing two transmembrane domains and a large NTPDase active, extracellular region containing apyrase conserved regions. It is heavily glycosolated, having six known N-glycosolation sites, and functions on the plasma membrane’s outer surface. CD39 initiates the metabolism, of ATP and ADP to adenosine through a 2-step exzymatic reaction, providing extracellular adenosine for the purpose of regulating various inflammatory endpoints. It is a putative marker for B-cell activation and its expression has also been observed on NK-cells, dendritic cells, T-cells, and vascular endothelial cells. CD39 is a known inhibitor of ADP-induced platelet aggregation and may also play a role in angiogenesis.

Specificity

This antibody recognizes CD39.

Immunogen

Recombinant protein corresponding to mouse CD39.

Application

This Anti-CD39 Antibody, clone 5F2 is validated for use in FC, WB for the detection of CD39.
Western Blot Analysis: A previous lot was used by an independent laboratory in WB.

Quality

Evaluated by Flow Cytometry in BALB/c splenocytes.

Flow Cytometry Analysis: 0.1 µg/mL of this antibody detected CD39 in BALB/c splenocytes.

Target description

~ 100 kDa
It has been published that CD39 shows molecular weights between ~70-100 kDa (Wang, T.F. et al., 1996).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service