Skip to Content
Merck
All Photos(1)

Key Documents

06-288

Sigma-Aldrich

Anti-Nck Antibody

Upstate®, from rabbit

Synonym(s):

Anti-NCK, Anti-nck-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€515.00

€515.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€515.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€515.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NCK1(4690)

Specificity

Nck

Immunogen

Purified GST fusion protein produced in E. coli., corresponding to human Nck amino acid sequence 1 to 254

Application

Detect Nck using this Anti-Nck Antibody validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated by immunoblot on human A431 carcinoma cells, mouse 3T3/A31 fibroblasts or rat L6 skeletal fibroblasts

Target description

47kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing 0.05% sodium azide
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Lock et al.
The Journal of biological chemistry, 274(32), 22775-22784 (1999-07-31)
A murine embryonic cDNA library was screened for potential substrates of the Src family kinase, Lyn, using a phosphorylation-screening strategy. One cDNA that we identified encodes Dok-related protein (DokR), a protein with homology to p62(dok) (Dok), and members of the
Kazuya Mizuno et al.
Journal of immunology (Baltimore, Md. : 1950), 169(2), 778-786 (2002-07-05)
Src homology region 2 domain-containing phosphatase 1 (SHP-1) is a key mediator in lymphocyte differentiation, proliferation, and activation. We previously showed that B cell linker protein (BLNK) is a physiological substrate of SHP-1 and that B cell receptor (BCR)-induced activation
Stanley S Stylli et al.
Journal of cell science, 122(Pt 15), 2727-2740 (2009-07-15)
Invadopodia are actin-based projections enriched with proteases, which invasive cancer cells use to degrade the extracellular matrix (ECM). The Phox homology (PX)-Src homology (SH)3 domain adaptor protein Tks5 (also known as SH3PXD2A) cooperates with Src tyrosine kinase to promote invadopodia
Angika Basant et al.
Microbiology spectrum, 11(6), e0152923-e0152923 (2023-10-19)
Vaccinia virus is a large double-stranded DNA virus and a close relative of Mpox and Variola virus, the causative agent of smallpox. During infection, Vaccinia hijacks its host's transport systems and promotes its spread into neighboring cells by recruiting a
João V Cordeiro et al.
PloS one, 4(12), e8506-e8506 (2009-12-31)
The cortical actin cytoskeleton beneath the plasma membrane represents a physical barrier that vaccinia virus has to overcome during its exit from an infected cell. Previous observations using overexpression and pharmacological approaches suggest that vaccinia enhances its release by modulating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service