Skip to Content
Merck
All Photos(3)

Key Documents

05-801

Sigma-Aldrich

Anti-trimethyl-Histone H3 (Lys36) Antibody, clone MC86, rabbit monoclonal

clone MC86, Upstate®, from rabbit

Synonym(s):

H3K36me3, Histone H3 (tri methyl K36)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€614.00

€614.00


Estimated to ship on22 March 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€614.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€614.00


Estimated to ship on22 March 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MC86, monoclonal

species reactivity

chicken

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
dot blot: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys36)

Specificity

Broad species cross-reactivity is expected.
Recognizes Histone H3 trimethylated on Lysine 36.

Immunogen

peptide containing the sequence GVme3KKP, in which me3K corresponds to trimethyl lysine 36 of human histone H3

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-trimethyl-Histone H3 (Lys36) Antibody, clone MC86 (rabbit monoclonal antibody) validated in DB, WB, PIA to detect trimethyl-Histone H3 (Lys36) also known as H3K36me3, Histone H3 (tri methyl K36).

Quality

routinely evaluated by immunoblot on chicken Core Histones (Catalog #13-107); did not detect unmodified recombinant Histone H3 (Catalog #14-411)

Target description

~17 kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin in a solution containing 0.07M Tris-glycine, 0.105M NaCl, pH 7.4, 0.035% sodium azide, 30% glycerol.
Protein A purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Acid extracted proteins from HeLa cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Zhang et al.
The Plant journal : for cell and molecular biology, 64(4), 604-617 (2010-09-09)
MADS-box transcription factors are known for their roles in plant growth and development. The regulatory mechanisms of spatial and temporal specific expression of MADS-box genes and the function of MADS-box genes in other biological processes are still to be explored.
The Iws1:Spt6:CTD complex controls cotranscriptional mRNA biosynthesis and HYPB/Setd2-mediated histone H3K36 methylation.
Yoh, SM; Lucas, JS; Jones, KA
Genes & Development null
Li-Qun Chen et al.
Plant physiology, 174(3), 1795-1806 (2017-05-28)
Methylation of Lys residues in the tail of the H3 histone is a key regulator of chromatin state and gene expression, conferred by a large family of enzymes containing an evolutionarily conserved SET domain. One of the main types of
Mass spectrometry analysis of the variants of histone H3 and H4 of soybean and their post-translational modifications.
Wu, T; Yuan, T; Tsai, SN; Wang, C; Sun, SM; Lam, HM; Ngai, SM
BMC plant biology null
Guochun Zhu et al.
International journal of biological sciences, 18(14), 5522-5538 (2022-09-24)
Cathepsins play a role in regulation of cell function through their presence in the cell nucleus. However, the role of Cathepsin K (Ctsk) as an epigenetic regulator in osteoclasts remains unknown. Our data demonstrated that Ctsk-/-Mmp9-/- mice have a striking

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service