Skip to Content
Merck

Skip To

04-790

Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30, rabbit monoclonal

culture supernatant, clone AW30, Upstate®

Synonym(s):

H3K4me2, Histone H3 (di methyl K4)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
AW30, monoclonal
Species reactivity:
chicken, human
Application:
ChIP, DB, WB, inhibition assay
Citations:
12
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AW30, monoclonal

species reactivity

chicken, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq), dot blot: suitable, inhibition assay: suitable (peptide), western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

dimethylation (Lys4)

Gene Information

human ... H3C1(8350)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Di- and trimethylation of histone H3 at Lys4 correlates with transcriptional activity of many genes. Dimethylation of Histone H3 at Lys4 occurs at both active and inactive euchromatic regions but not in silent heterochromatic sites, whereas trimethylation at Lys4 is present exclusively at active genes.
~17kDa

Immunogen

KLH-conjugated, synthetic peptide containing the sequence …Tme2KQT… in which me2K corresponds to dimethyl-lysine at residue 4 of human histone H3.

Application

Western Blot Analysis:
A 1:1000-1:2000 dilution of this lot detected dimethyl histone H3 (Lys4) in HeLa acid extracts.

Western Blot Analysis:
A 1:2,000 to 1:10,000 dilution of a previous lot detected methylated histone H3 in acid extracted proteins from HeLa cells. The antibody did not detect unmethylated recombinant Histone H3.

ChIP-seq Analysis:
Representative lot data. Chromatin immunoprecipitation was performed using the Magna ChIP HiSens kit (cat# 17-10460), Anti-dimethyl-Histone H3 (Lys4) antibody (2 µl cat# 04-790), 20 µL Protein A/G beads, and 1e6 crosslinked HeLa cell chromatin followed by DNA purification using magnetic beads. Libraries were prepared from Input and ChIP DNA samples using standard protocols with Illumina barcoded adapters, and analyzed on Illumina HiSeq instrument. An excess of sixteen million reads from FastQ files were mapped using Bowtie (http://bowtie-bio.sourceforge.net/manual.shtml) following TagDust (http://genome.gsc.riken.jp/osc/english/dataresource/) tag removal. Peaks were identified using MACS (http://luelab.dfci.harvard.edu/MACS/), with peaks and reads visualized as a custom track in UCSC Genome Browser (http://genome.ucsc.edu) from BigWig and BED files. The highest 25% of peaks identified in the 04-790 and 05-1338 datasets showed 92 and 90% overlap with peaks identified in the ENCODE H3K4me2 BROAD Histone track for HeLa S3.

Dot Blot Analysis:
Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with Cat. No 04-790, Anti-dimethyl Histone H3 (Lys4), clone AW30 at 1:1000 dilution. Proteins were visualized using a Donkey anti-Rabbit IgG conjugated to HRP and a chemiluminescence detection system.

Peptide Inhibition Analysis:
2 μM of a histone H3 peptide containing dimethyl-lysine 4 abolished detection of histone H3 by a previous lot of antibody in immunoblot analysis of HeLa acid extracts. No signal reduction was observed with histone H3 peptides containing either monomethyl or trimethyl-lysine 4 modifications
Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30 is a rabbit monoclonal antibody for detection of dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4) & has been validated in WB, ChIP, ChIP-seq, DB, PIA, Mplex.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Biochem/physiol Actions

Histone H3 dimethylated on lysine 4
The immunizing sequence is conserved from Tetrahymena to human, so broad species cross-reactivity is expected.

Physical form

Cultured supernantant in 0.05% sodium azide

Preparation Note

Stable for 1 year at -20°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
HeLa acid extracts
routinely evaluated by immunoblot on acid-extracted proteins from HeLa cells; the antibody did not detect unmethylated recombinant Histone H3 (Catalog #14-494)

Other Notes

Replaces: 05-790

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


WGK

WGK 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters