Skip to Content
Merck
All Photos(1)

Key Documents

05-513

Sigma-Aldrich

Anti-ATM Antibody, clone AM9

ascites fluid, clone AM9, Upstate®

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€358.00

€358.00


Estimated to ship on20 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1376

Request a Bulk Order

Select a Size

Change View
200 μL
€358.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€358.00


Estimated to ship on20 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1376

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

AM9, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ATM(472)

Specificity

ATM

Immunogen

Full-length human ATM

Application

Detect ATM also known as Ataxia Telangiectasia Mutated with Anti-ATM Antibody, clone AM9 (Mouse Monoclonal Antibody), that has been demonstrated to work in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot on nuclear extract from Raji cells

Target description

350kDa

Linkage

Replaces: 04-200

Physical form

Ascites
mouse ascites containing 0.05% sodium azide

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Wang et al.
Translational oncology, 10(2), 190-196 (2017-02-10)
The ataxia telangiectasia mutated (ATM) protein kinase plays a central role in the cellular response to DNA damage. Loss or inactivation of both copies of the ATM gene (ATM) leads to ataxia telangiectasia, a devastating childhood condition characterized by neurodegeneration
T K Pandita et al.
Molecular and cellular biology, 19(7), 5096-5105 (1999-06-22)
A-T (ataxia telangiectasia) individuals frequently display gonadal atrophy, and Atm-/- mice show spermatogenic failure due to arrest at prophase of meiosis I. Chromosomal movements take place during meiotic prophase, with telomeres congregating on the nuclear envelope to transiently form a
Scott L Baughan et al.
Cancer genetics, 264-265, 40-49 (2022-03-31)
Variants of unknown significance (VUS) remain a constant challenge in the diagnosis of hereditary cancer and the counseling of patients with pedigrees suggestive of such a syndrome. In order to assess some of this limitation, several variants in the DNA
K Nakagawa et al.
Molecular and cellular biology, 19(4), 2828-2834 (1999-03-19)
Microinjection of the restriction endonuclease HaeIII, which causes DNA double-strand breaks with blunt ends, induces nuclear accumulation of p53 protein in normal and xeroderma pigmentosum (XP) primary fibroblasts. In contrast, this induction of p53 accumulation is not observed in ataxia
Characterization of mre11 loss following HSV-1 infection.
Gregory, DA; Bachenheimer, SL
Virology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service