Skip to Content
Merck
All Photos(1)

Key Documents

05-406

Sigma-Aldrich

Anti-JAK3 Antibody, clone B32-32

clone B32-32, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€347.00

€347.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€347.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€347.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

B32-32, monoclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... JAK3(3718)
mouse ... Jak3(16453)

Specificity

JAK3

Immunogen

GST-fusion protein containing amino acids 1076-1100 of murine thymus JAK3. Clone B32-32.

Application

Anti-JAK3 Antibody, clone B32-32 detects level of JAK3 & has been published & validated for use in IP.
Not recommended for WB.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

routinely evaluated by immunoprecipitation of JAK3 from CTLL cells or from TF-1 RIPA cell lysate

Target description

125kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing 0.05% sodium azide
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T L Wong et al.
Journal of applied microbiology, 97(6), 1185-1191 (2004-11-18)
To obtain preliminary data on the microbiology and hurdles to pathogen growth in the traditional Pacific Island food, povi masima, which is essentially beef brisket cured in brine. Six containers of povi masima were prepared and two were inoculated with
C C Tassou et al.
Journal of applied microbiology, 104(6), 1764-1773 (2008-02-27)
The purpose of this study was to investigate the inactivation kinetics of Staphylococcus aureus in a ham model system by high hydrostatic pressure at ambient (25 degrees C) and selected temperatures (45, 55 degrees C). Selective [Baird Parker (BP) agar]
JAK-mediated phosphorylation and activation of STAT signaling proteins. Analysis by phosphotyrosine blotting and EMSA.
N Broughton et al.
Methods in molecular biology (Clifton, N.J.), 124, 131-146 (2000-12-02)
C B Gurniak et al.
Oncogene, 13(4), 777-786 (1996-08-15)
Using a PCR-based screen to identify tyrosine kinases involved in T cell development, we have cloned a new member of the Eph-family of receptor tyrosine kinases (Mep, for murine eph-family protein). At the amino acid level Mep is 60% identical
S Ogur
Brazilian journal of biology = Revista brasleira de biologia, 82, e262735-e262735 (2022-07-07)
This study was conducted to determine the pathogenic bacteria load of 14 species of marine fish obtained from two suppliers (in Bitlis city, Turkey), which provide fish for fish markets, and to reveal the safety of the marine fish in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service