Skip to Content
Merck
All Photos(1)

Key Documents

05-180

Sigma-Aldrich

Anti-Cortactin (p80/85) Antibody, clone 4F11

clone 4F11, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

4F11, monoclonal

species reactivity

mouse, bovine, hamster, human, rat, avian

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CTTN(2017)

General description

Cortactin is present as a p80 and p85 kDa protein in many cell types. The protein was first identified as being a major substrate of the tyrosine kinase pp60SRC. The main structural feature of the protein is that at the N-terminus there are five and a half 37-amino acid repeats, these appear to be responsible for the actin-binding. This repeat region occupies about a third of the protein. The repeat region is followed by a helical region and then a proline rich region and finally at the C-terminus, an SH3 domain. The proline rich domain is thought to bind the SH2 domain of v-src the viral version of the normal c-src known to phosphorylate cortactin. Cortactin bundles microfilaments and this bundling is inhibited by tyrosine phosphorylation. One pathway that activates the pathway leading to tyrosine phosphorylation of cortactin results from a cell adhesion molecule in brain micro-vessel endothelial cells, another is the growth factor, but presumably the very many mechanisms for pp60src activation would also result in cortactin phosphorylation and subsequent loss of actin binding.

Specificity

Recognizes Cortactin.

Immunogen

Mixture of affinity-purified tyrosine phosphoproteins from chick embryo fibroblasts expressing activated pp60src

Application

Anti-Cortactin (p80/85) Antibody, clone 4F11 is an antibody against Cortactin (p80/85), and has been published and validated for use in IC, IH, IP and WB.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

routinely evaluated in mouse 3T3/A31 cell lysates or chicken fibroblast src-transformed cell lysate

Target description

80 kDa and 85 kDa

Physical form

Format: Purified
Protein G Purified
Purified immunoglobulin in 10mM PBS, pH 7.4 with 0.05% sodium azide.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Control Included: 3T3 Cell Lysate (12-305) Other controls: PC-12 cell lysate, HeLa whole cell lysate or A-431 whole cell lysate

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuba stimulates intracellular N-WASP-dependent actin assembly.
Kovacs, EM; Makar, RS; Gertler, FB
Journal of Cell Science null
Inactivation of Src family kinases inhibits angiogenesis in vivo: implications for a mechanism involving organization of the actin cytoskeleton.
Witold W Kilarski, Natalia Jura, Par Gerwins
Experimental Cell Research null
Direct interaction between caldesmon and cortactin.
Renjian Huang, Gong-Jie Cao, Hongqiu Guo, Jolanta Kordowska, C-L Albert Wang
Archives of Biochemistry and Biophysics null
Xiaolin Sun et al.
Carcinogenesis, 30(12), 2109-2116 (2009-10-22)
Abl interactor 1 (Abi1) is a key regulator of actin polymerization/depolymerization. The involvement of Abi1 in the development of abnormal cytoskeletal functions of cancer cells has recently been reported. It remains unclear, however, how Abi1 exerts its effects in tumor
Hippocampal Cortactin Levels are Reduced Following Spatial Working Memory Formation, an Effect Blocked by Chronic Calpain Inhibition.
Olson, ML; Ingebretson, AE; Harmelink, KM
Brain sciences null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service