Skip to Content
Merck
All Photos(1)

Documents

09-881

Sigma-Aldrich

Anti-acetyl Cortactin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Amplaxin, Oncogene EMS1, Src substrate cortactin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

Drosophila (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (not specified)

Gene Information

human ... CTTN(2017)

General description

Cortactin is a cytoskeleton protein that facilitates assembly of cortical actin. Cortactin is widely expressed in most adherent cells and is a prominent substrate of protein-tyrosine kinase Src in vivo and in vitro. The protein sequence of cortactin contains multiple amino-terminal tandem repeats of a unique 37-amino acid sequence, which associates with actin, and a Src homology 3 domain at the carboxyl terminus.

Recent research efforts show cortactin is acetylated in vivo and is deacetylated by HDAC6 (Histone deacetylase 6). HDAC6 is a tubulin-specific deacetylase that regulates microtubule-dependent cell movement. In recent studies it was demonstrated that HDAC6 binds cortactin and alters the ability of cortactin to bind F-actin by modulating a “charge patch” in its repeat region. Findings suggest that, in addition to its role in microtubule-dependent cell motility, HDAC6 influences actin-dependent cell motility by altering the acetylation status of cortactin, which, in turn, changes the F-actin binding activity of cortactin. The acetylation level of cortactin may correlate to tumor metastases and acetylated cortactin may be considered a potential metastasis marker.

Specificity

The antibody recognizes human acetylated Cortactin.

Immunogen

Consensus sequence motif of acetylation sites of cortactin.
Epitope: Unknown

Application

Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-acetyl Cortactin Antibody is validated for use in Western Blotting, for the detection of acetyl Cortactin.

Quality

Evaluated by Western Blot in HeLa-Anti-Cortactin immunocomplex treated with TSA/Nicotinamide.

Western Blot Analysis: 2.5 µg/ml of this antibody detected Cortactin on 10 µL of HeLa-Anti-Cortactin immunocomplex treated with TSA/Nicotinamide.

Target description

~ 85 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa-Anti-Cortactin immunocomplex treated with TSA/Nicotinamide

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongying Mo et al.
Nature communications, 9(1), 1007-1007 (2018-03-10)
Dominant mutations in glycyl-tRNA synthetase (GlyRS) cause a subtype of Charcot-Marie-Tooth neuropathy (CMT2D). Although previous studies have shown that GlyRS mutants aberrantly interact with Nrp1, giving insight into the disease's specific effects on motor neurons, these cannot explain length-dependent axonal
Pedro Guedes-Dias et al.
Biochimica et biophysica acta, 1852(11), 2484-2493 (2015-08-25)
Striatal neurons are vulnerable to Huntington's disease (HD). Decreased levels of acetylated alpha-tubulin and impaired mitochondrial dynamics, such as reduced motility and excessive fission, are associated with HD; however, it remains unclear whether and how these factors might contribute to
Vugar Azizov et al.
iScience, 26(7), 107230-107230 (2023-07-24)
Alcohol is among the most widely consumed dietary substances. Excessive alcohol consumption damages the liver, heart, and brain. Alcohol also has strong immunoregulatory properties. Here, we report how alcohol impairs T cell function via acetylation of cortactin, a protein that binds
Eugenia Meiler et al.
PloS one, 7(3), e33662-e33662 (2012-04-06)
Cortactin is a classical Src kinase substrate that participates in actin cytoskeletal dynamics by activating the Arp2/3 complex and interacting with other regulatory proteins, including FAK. Cortactin has various domains that may contribute to the assembly of different protein platforms
Joana Margarida Silva et al.
Cell death and differentiation, 26(8), 1411-1427 (2018-11-18)
Imbalance of neuronal proteostasis associated with misfolding and aggregation of Tau protein is a common neurodegenerative feature in Alzheimer's disease (AD) and other Tauopathies. Consistent with suggestions that lifetime stress may be an important AD precipitating factor, we previously reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service