Skip to Content
Merck
All Photos(1)

Documents

05-1572

Sigma-Aldrich

Anti-MDC1 Antibody, clone P2B11

clone P2B11, from mouse

Synonym(s):

Nuclear factor with BRCT domains 1, homologue to Drosophila photoreceptor protein calphotin, mediator of DNA damage checkpoint 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P2B11, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), bovine (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MDC1(9656)
mouse ... Mdc1(240087)

General description

MDC1 is a subunit of the cohesin complex and is an essential protein required for sister chromatid cohesion in mitosis and meiosis. Temperature-sensitive mutations of this protein have been associated with defects in sister chromatid cohesion, chromosome condensation, and chromosome segregation. Its expression is cell cycle regulated and peaks in S phase. Expression of MDC1 is required for the S- and G2/M-checkpoint-mediated cell cycle arrest in response to DNA damage. MDC1 has been shown to interact with H2A.X near sites of DNA double-strand breaks and recruit ATM kinase to the damaged region of DNA. Consistent with this role, MDC1 also appears to modulate the function of other proteins involved in the DNA damage checkpoint such as BRCA1 and Chk2.

Specificity

This antibody recognizes MDC1 at and around the N-terminus.

Immunogen

GST-tagged recombinant protein corresponding to mouse MDC1 at and around the N-terminus.

Application

Anti-MDC1 Antibody, clone P2B11 is a Mouse Monoclonal Antibody for detection of MDC1 also known as Nuclear factor with BRCT domains 1 & has been validated in ICC & WB.
Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected MDC1 in NIH/3T3 cells.

Western Blot Analysis: A representative lot was used by an independent laboratory in WB. (Lou, Z., et al. (2003). Nature. 421(6926):957-961.)

Target description

~ 184 kDa There are 4 isoforms of human MDC1 because of splicing with molecular weights of approximately 227, 198, 196, and 117 kDa

Physical form

Format: Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) without 0.05% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Mayer et al.
Cell cycle (Georgetown, Tex.), 15(4), 546-558 (2016-01-09)
Because low levels of DNA double strand breaks (DSBs) appear not to activate the ATM-mediated prophase I checkpoint in full-grown oocytes, there may exist mechanisms to protect chromosome integrity during meiotic maturation. Using live imaging we demonstrate that low levels
Sahra Bodo et al.
The Journal of clinical investigation, 129(2), 786-801 (2018-11-28)
Tumor cure with conventional fractionated radiotherapy is 65%, dependent on tumor cell-autonomous gradual buildup of DNA double-strand break (DSB) misrepair. Here we report that single-dose radiotherapy (SDRT), a disruptive technique that ablates more than 90% of human cancers, operates a
Kuntian Luo et al.
Genes & development, 30(23), 2581-2595 (2016-12-13)
Homologous recombination (HR) is one of the major DNA double-strand break (DSB) repair pathways in mammalian cells. Defects in HR trigger genomic instability and result in cancer predisposition. The defining step of HR is homologous strand exchange directed by the
Hung-Sheng Shang et al.
Oncology reports, 36(4), 2207-2215 (2016-08-09)
Cervical cancer is one of the most common cancers in women worldwide and it is a prominent cause of cancer mortality. Curcumin is one of the major compounds from Turmeric and has been shown to induce cytotoxic cell death in
Yung-Luen Shih et al.
Oncology reports, 36(4), 2094-2100 (2016-08-31)
Casticin, a polymethoxyflavone, has been demonstrated to possess anticancer activities, yet no study has shown in detail that casticin induces DNA damage in lung cancer cells. The purpose of this study was to investigate the possible molecular mechanisms of casticin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service