Skip to Content
Merck
All Photos(1)

Key Documents

T-108

Supelco

THC Cannabinoids Mixture-3 solution

1.0 mg/mL in methanol (each analyte), certified reference material, ampule of 0.5 mL, Cerilliant®

Synonym(s):

THC and Cannabinoid Standard Mix

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O2
Molecular Weight:
310.43
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®]]

composition

Cannabidiol, 1.0 mg/mL
Cannabinol, 1.0 mg/mL
(-)-delta9-THC, 1.0 mg/mL

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (each analyte)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
forensics and toxicology

format

multi-component solution

storage temp.

−20°C

General description

A multi-component certified solution standard of (-)-Δ9-THC, cannabinol (CBN) and cannabidiol (CBD). This multi-component mix is suitable for cannabinoid testing methods by GC/MS, LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling, pharmaceutical research, and forensic analysis. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

HPLC separates vitamin E acetate, THC, and CBD in vape products, providing accurate analysis of commercially available vape liquids.

Cannabis products need cannabinoid characterization. Ascentis Express C18 resolves 14 similar cannabinoids in under seven minutes with excellent peak shape.

ISO-certified cannabinoid CRMs, THC and THCA, available as single components or mixes, for accurate potency testing.

Protocols

Cannabis products need cannabinoid characterization. Ascentis Express C18 resolves 14 similar cannabinoids in under seven minutes with excellent peak shape.

Questions

  1. Is a State license required to purchase cannabis standards?

    1 answer
    1. This product is a mixture of components in methanol, as are most of the cannabis standards. The presence of methanol eliminates the possibility of misuse. Standards prepared in solutions of methanol or other organic solvents do not require a state license in order to purchase.

      Please navigate to the link https://www.sigmaaldrich.com/US/en/support/customer-support, click on "Products
      Pricing & Availability" under the Products Section with all the required information so that a member of the Customer Service team can reach out to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service