Skip to Content
Merck
All Photos(1)

Key Documents

T-076

Supelco

L-Thyroxine-13C6 solution

100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€257.00

€257.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€257.00

About This Item

Empirical Formula (Hill Notation):
13C6C9H11I4NO4
CAS Number:
Molecular Weight:
782.83
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€257.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 0.1N NH3

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

I[13C]1=[13C]([13C](I)=[13CH][13C](C[C@@H](C(O)=O)N)=[13CH]1)OC2=CC(I)=C(O)C(I)=C2

InChI

1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1/i1+1,2+1,6+1,10+1,11+1,14+1

InChI key

XUIIKFGFIJCVMT-PDHVKVOFSA-N

General description

This internal standard and Certified Spiking Solution® is suitable for quantitation of free and total T4 serum levels by LC-MS/MS for determination of hyper- or hypothyroidism and monitoring treatment with synthetic thyroid hormones.

L-Thyroxine-13C6 is an isotopically labeled form of the tyrosine-based thyroid hormone, thyroxine (T4). Thyroxine is secreted by the thyroid gland and is involved in different biological processes, such as growth, neurodevelopment, carbohydrate & oxygen metabolism, and protein synthesis

Application

The Certified Reference Material (CRM) in solution can also be used as an internal standard as follows:
  • Combination of hollow fiber-liquid-phase microextraction (HF-LPME) based sample preparation and liquid chromatography-tandem mass spectrometric (LC-MS/MS) determination of total thyroxine in serum samples
  • Development of an LC-MS/MS method for the multi-residue analysis of melatonin, cortisol, triiodothyronine, thyroxine, and testosterone in rat serum samples following their dual liquid-liquid extraction
  • Simultaneous determination of thyroxine by an isotope dilution-matrix-assisted laser desorption ionization-time of flight mass spectrometry (MALDI-TOF MS) method from human serum samples
  • Estimation of thyroxine (T4) and triiodothyronine (T3) in rat cardiac tissue samples by a high-performance liquid chromatographic-tandem mass spectrometric (HPLC-MS/MS) method following their derivatization
  • Establishing and validating an LC-MS/MS-based method for the quantification of endogenous thyroid hormones, T4 and two of its T3 isomers, in tissue samples obtained from lamprey, following the enzymatic digestion and protein precipitation of the samples

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Determination of free thyroid hormones (T3 & T4) in serum by LC-MS/MS following a streamlined 1-hour BioSPME sample preparation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service