Skip to Content
Merck
All Photos(1)

Key Documents

H-059

Supelco

6β-Hydroxytestosterone solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€100.00

€100.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€100.00

About This Item

Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€100.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H]3[C@@H](C[C@@H](O)C4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H28O3/c1-18-7-5-11(20)9-15(18)16(21)10-12-13-3-4-17(22)19(13,2)8-6-14(12)18/h9,12-14,16-17,21-22H,3-8,10H2,1-2H3/t12-,13-,14-,16+,17-,18+,19-/m0/s1

InChI key

XSEGWEUVSZRCBC-ZVBLRVHNSA-N

General description

A Certified Spiking Solution® suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical and diagnostic testing to endocrinology and sports testing. 6ß-Hydroxytestosterone is a major urinary metabolite of testosterone, a hormone from the androgen group of steroids. Testosterone is an important sex hormone and anabolic steroid used as a performance-enhancing drug.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Liu et al.
Biological & pharmaceutical bulletin, 27(10), 1555-1560 (2004-10-07)
The intestinal bacterial metabolites of ginsenosides are responsible for the main pharmacological activities of ginseng. The purpose of this study was to find whether these metabolites influence hepatic metabolic enzymes and to predict the potential for ginseng-prescription drug interactions. Utilizing
Cuiping Chen et al.
The Journal of pharmacology and experimental therapeutics, 300(2), 417-420 (2002-01-24)
The adrenochrome reaction (oxidation of epinephrine to adrenochrome) has been widely employed as a standard assay for reactive oxygen species, produced under a variety of conditions, including those produced during cytochrome P450 (CYP)-mediated oxidation of substrates such as cyclosporine. However
Dan A Rock et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(12), 2410-2413 (2008-09-04)
Cytochrome P450 (P450) reaction phenotyping is a key process toward accurately determining the contribution of different P450s to the metabolism of new chemical entities. The significance of P450s to drug disposition has led to the identification of selective chemical and
Thomas M Polasek et al.
European journal of clinical pharmacology, 62(3), 203-208 (2006-01-18)
Macrolide antibiotics are mechanism-based inactivators of CYP3A enzymes that exhibit varying degrees of inhibitory potency. Our aim was to predict quantitatively the drug-drug interaction (DDI) potential of five macrolides from in vitro studies using testosterone as the CYP3A substrate, and
Snezana Mirkov et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(2), 228-233 (2006-11-17)
The effects of green tea compounds on the metabolism of irinotecan have never been investigated. We aimed to study whether catechins [(-)-epigallocatechin gallate (EGCG), (-)-epicatechin gallate (ECG), (-)-epigallocatechin (EGC), (-)-epicatechin] affect the inactivation metabolism of irinotecan into 7-ethyl-10-[4-N-(1-piperidino)-1-amino]carbonyloxycamptothecin (NPC) (by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service