Skip to Content
Merck
All Photos(1)

Key Documents

P-111

Supelco

Promethazine hydrochloride solution

1.0 mg/mL (1% 1 M HCl in Methanol (as free base)), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€69.30

€69.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€69.30

About This Item

Empirical Formula (Hill Notation):
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€69.30


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL (1% 1 M HCl in Methanol (as free base))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

S1c2c(cccc2)N(c3c1cccc3)CC(N(C)C)C.[Cl-].[H+]

InChI

1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H

InChI key

XXPDBLUZJRXNNZ-UHFFFAOYSA-N

General description

Promethazine is a phenothiazine analog prescribed for treatment of nausea, motion sickness, vomiting, and allergies. Sold under trade names such as Phenergan® and Phenadoz®, promethazine has antihistamine, sedative, and mild antipsychotic effects. This Certified Spiking Solution® is suitable for GC/MS or LC/MS methods in promethazine testing applications including clinical toxicology, forensic analysis, and urine drug testing. Among drug abusers, promethazine has reportedly been misused in conjunction with opioids, benzodiazepines, and sedatives.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Phenadoz is a registered trademark of Watson Laboratories, Inc.
Phenergan is a registered trademark of Delcor Asset Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service