Skip to Content
Merck
All Photos(1)

Key Documents

N-033

Supelco

Noroxycodone-D3 hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Labeled Noroxycodone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16D3NO4·HCl
Molecular Weight:
340.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.[2H]C([2H])([2H])Oc1ccc2C[C@H]3NCC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O

InChI

1S/C17H19NO4.ClH/c1-21-11-3-2-9-8-12-17(20)5-4-10(19)15-16(17,6-7-18-12)13(9)14(11)22-15;/h2-3,12,15,18,20H,4-8H2,1H3;1H/t12-,15+,16+,17-;/m1./s1/i1D3;

InChI key

IGNAMRAQFUFUMH-HHAOZYOFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This stable-labeled certified reference solution is suitable for quantitation of noroxycodone levels in urine, serum, or plasma by LC/MS or GC/MS for applications such as urine drug testing, clinical toxicology, forensic analysis, pain prescription monitoring, or isotope dilution methods. Noroxycodone is a major urinary metabolite of oxycodone, an opiate analgesic drug used for the management of moderate to severe pain, both acute and chronic.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service