Skip to Content
Merck
All Photos(1)

Key Documents

M-182

Supelco

Mitragynine-D3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€268.00

€268.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€268.00

About This Item

Empirical Formula (Hill Notation):
C23H27D3N2O4
Molecular Weight:
401.51
UNSPSC Code:
41116107
NACRES:
NA.24

€268.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−70°C

General description

An internal standard suitable for quantitation of mitragynine in biological samples using mass spectrometry-based methods. Mitragynine, commonly known as Kratom, is the most abundant alkaloid in the plant Mitragynine speciosa. Sold over the internet as herbal plant extracts, Kratom is noted for its psychoactive, stimulant, and sedative effects depending on the dosage level. According to DEA′s Drugs and Chemicals of Concern page, mitragynine use can lead to addiction with adverse effects ranging from anorexia and weight loss to hallucinations and delusion.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service