Skip to Content
Merck
All Photos(1)

Key Documents

M-152S

Supelco

Mitragynine solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Mitragynine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N2O4
CAS Number:
Molecular Weight:
398.50
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

-10 to -25°C

SMILES string

O=C(OC)/C([C@H]1C[C@@](N(C[C@H]1CC)CC2)([H])C3=C2C(C(OC)=CC=C4)=C4N3)=C/OC

InChI

1S/C23H30N2O4/c1-5-14-12-25-10-9-15-21-18(7-6-8-20(21)28-3)24-22(15)19(25)11-16(14)17(13-27-2)23(26)29-4/h6-8,13-14,16,19,24H,5,9-12H2,1-4H3/b17-13+/t14-,16+,19+/m1/s1

InChI key

LELBFTMXCIIKKX-QVRQZEMUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mitragynine, commonly known as Kratom, is the most abundant alkaloid in the plant Mitragynine speciosa. Sold over the internet as herbal plant extracts, Kratom is noted for its psychoactive, stimulant, and sedative effects depending on the dosage level. According to DEA′s Drugs and Chemicals of Concern page, mitragynine use can lead to addiction with adverse effects ranging from anorexia and weight loss to hallucinations and delusion.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service