Skip to Content
Merck
All Photos(1)

Key Documents

H-092

Supelco

4-Hydroxy-3-methoxyphenyl-D3-acetic-D2 Acid solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

4-Hydroxy-3-methoxyphenyl-d3-acetic-d2 Acid (HVA-d5), HVA-D5 solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€166.00

€166.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€166.00

About This Item

Empirical Formula (Hill Notation):
C9H5D5O4
CAS Number:
Molecular Weight:
187.20
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€166.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O(C)c1c(c(c(c(c1[2H])C(C(=O)O)([2H])[2H])[2H])[2H])O

InChI

1S/C9H10O4/c1-13-8-4-6(5-9(11)12)2-3-7(8)10/h2-4,10H,5H2,1H3,(H,11,12)/i2D,3D,4D,5D2

InChI key

QRMZSPFSDQBLIX-DVHRWRHBSA-N

General description

Homovanillic acid (HVA) is a urinary metabolite of the catecholamine dopamine and is associated with dopamine levels in the brain. This stable labeled internal standard is suitable for quantitating HVA levels in LC/MS applications such as clinical and diagnostic testing or endocrinology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijie Ouyang et al.
Investigative ophthalmology & visual science, 62(1), 25-25 (2021-01-26)
To evaluate the role of CD4+ T helper cells in benzalkonium chloride (BAC)-induced ocular surface disorder in C57BL/6 mice. Topical 0.075% BAC was applied twice daily in C57BL/6 mice for 7 consecutive days; PBS-treated and untreated mice served as controls.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service