Skip to Content
Merck
All Photos(1)

Key Documents

LM1504

Avanti

17:0-14:1 PI

Avanti Research - A Croda Brand LM1504, methanol solution

Synonym(s):

1-heptadecanoyl-2-(9Z-tetradecenoyl)-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€161.00

€161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€161.00

About This Item

Empirical Formula (Hill Notation):
C40H78NO13P
CAS Number:
Molecular Weight:
812.02
UNSPSC Code:
12352211
NACRES:
NA.25

€161.00


Check Cart for Availability

Request a Bulk Order

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1504-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1504

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(OC(CCCCCCC/C=C\CCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+]

InChI key

VDNRHLFKICVCHS-ATJCMGOSSA-N

Application

17:0-14:1 PI or 1-heptadecanoyl-2-(9Z-tetradecenoyl)-sn-glycero-3-phospho-(1′-myo-inositol) has been used as an internal standard for:
  • the detection of phosphatidylinositol class of lipids by tandem mass spectrometry (MS/MS) analysis[1]
  • phospholipid quantification by MS analysis[2]
  • lipid analysis of murine brain lipid extract using Fourier transform-ion cyclotron resonance-mass spectrometry (FT-ICR-MS)[3]

Packaging

2 mL Amber Glass Sealed Ampule (LM1504-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew W Granger et al.
Journal of neurochemistry, 149(4), 499-517 (2018-07-25)
Changes in glycerophosphocholine metabolism are observed in Alzheimer's disease; however, it is not known whether these metabolic disruptions are linked to cognitive decline. Here, using unbiased lipidomic approaches and direct biochemical assessments, we profiled Land's cycle lipid remodeling in the
Andreas Ullen et al.
Free radical biology & medicine, 49(11), 1655-1665 (2010-09-03)
Plasmalogens, 1-O-alk-1'-enyl-2-acyl-sn-glycerophospholipids, are significant constituents of cellular membranes and are essential for normal brain development. Plasmalogens, which contain a vinyl ether bond at the sn-1 position, are preferential targets for hypochlorous acid (HOCl), generated by myeloperoxidase (MPO) from H(2)O(2) and
Ulrike Bruning et al.
Cell metabolism, 28(6), 866-880 (2018-08-28)
The role of fatty acid synthesis in endothelial cells (ECs) remains incompletely characterized. We report that fatty acid synthase knockdown (FASNKD) in ECs impedes vessel sprouting by reducing proliferation. Endothelial loss of FASN impaired angiogenesis in vivo, while FASN blockade reduced
Yakir Guri et al.
Cancer cell, 32(6), 807-823 (2017-12-13)
Dysregulated mammalian target of rapamycin (mTOR) promotes cancer, but underlying mechanisms are poorly understood. We describe an mTOR-driven mouse model that displays hepatosteatosis progressing to hepatocellular carcinoma (HCC). Longitudinal proteomic, lipidomics, and metabolomic analyses revealed that hepatic mTORC2 promotes de

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service