Skip to Content
Merck
All Photos(2)

Key Documents

900603P

Avanti

PhotoClick GM1 (synthetic)

Avanti Research - A Croda Brand

Synonym(s):

PhotoClick GM1 Ganglioside (synthetic, ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€863.00

€863.00


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€863.00

About This Item

Empirical Formula (Hill Notation):
C70H122N6O31
CAS Number:
Molecular Weight:
1543.74
UNSPSC Code:
12161700
NACRES:
NA.25

€863.00


Estimated to ship on18 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

package of 1 × 1 mg
package of 1 × 500 μg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCC1(N=N1)CCCC#C)=O)CO[C@H](O2)[C@H](O)[C@@H](O)[C@H](O[C@@H]3O[C@@H]([C@@H]([C@H](O[C@@]4(C([O-])=O)O[C@@H]([C@H](O)[C@H](O)CO)[C@@](NC(C)=O)([H])[C@@H](O)C4)[C@H]3O)O[C@@H]5O[C@@H]([C@@H]([C@H](O[C@@H]6O

Packaging

5 mL Amber Glass Screw Cap Vial (900603P-1MG)
5 mL Amber Glass Screw Cap Vial (900603P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service