Skip to Content
Merck
All Photos(1)

Key Documents

860572P

Avanti

C17 Mono-Sulfo Galactosyl(β) Ceramide (d18:1/17:0)

Avanti Research - A Croda Brand 860572P, powder

Synonym(s):

3-O-sulfo-D-galactosyl-β1-1′-N-heptadecanoyl-D-erythro-sphingosine (ammonium salt), (synthetic)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€429.00

€429.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€429.00

About This Item

Empirical Formula (Hill Notation):
C41H82N2O11S
CAS Number:
Molecular Weight:
811.16
UNSPSC Code:
12352211
NACRES:
NA.25

€429.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860572P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860572P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](OS([O-])(=O)=O)[C@H]([C@H]1CO)O.[NH4+]

InChI

1S/C41H79NO11S.H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-37(45)42-34(35(44)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)33-51-41-39(47)40(53-54(48,49)50)38(46)36(32-43)52-41;/h28,30,34-36,38-41,43-44,46-47H,3-27,29,31-33H2,1-2H3,(H,42,45)(H,48,49,50);1H

InChI key

YMVAMOSEAIAACO-FDBYLCKRSA-N

Application

C17 Mono-Sulfo Galactosyl(β) Ceramide (d18:1/17:0) is suitable for use as one of the lipid standards for quantification of different phospholipids by flow injection analysis-tandem mass spectrometry (FIA-MS/MS) method.[1][2]

Biochem/physiol Actions

C17 Mono-Sulfo Galactosyl(β) Ceramide (d18:1/12:0) is a non-physical sulfatide. Accumulation of sulfatide impairs the activity of arylsulfatase A enzyme, which in turn leads to the development of genetic disease, metachromatic leukodystrophy.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860572P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic sulfogalactosylceramide (sulfatide) and its use for the mass spectrometric quantitative urinary determination in metachromatic leukodystrophies
Cui Y, et al.
Glycoconjugate Journal, 25(2), 147-155 (2008)
Extracellular vesicles released by fibroblasts undergoing H-Ras induced senescence show changes in lipid profile
Buratta S, et al.
PLoS ONE, 12(11), e0188840-e0188840 (2017)
Diabetes alters myelin lipid profile in rat cerebral cortex: Protective effects of dihydroprogesterone
Cermenati G, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 168, 60-70 (2017)
He Li et al.
Food chemistry, 310, 125947-125947 (2019-12-17)
An ultra performance liquid chromatography-Triple time of flight mass spectrometry (UPLC-Triple TOF-MS/MS) method were established to characterize the lipid profiles in four shellfish species. More than 600 lipid molecular species belonging to 14 classes were detected. Phospholipids (PLs) were predominant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service