Skip to Content
Merck
All Photos(1)

Key Documents

900151P

Avanti

CoQ8

Avanti Research - A Croda Brand

Synonym(s):

Coenzyme Q8 (E. coli)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€187.00

€187.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€187.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€187.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (900151P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C1C(OC)=C(OC)C(C(C)=C1C/C=C(C)/CCC=C(C)C)=O

InChI

1S/C49H74O4/c1-36(2)20-13-21-37(3)22-14-23-38(4)24-15-25-39(5)26-16-27-40(6)28-17-29-41(7)30-18-31-42(8)32-19-33-43(9)34-35-45-44(10)46(50)48(52-11)49(53-12)47(45)51/h20,22,24,26,28,30,32,34H,13-19,21,23,25,27,29,31,33,35H2,1-12H3/b37-22+,38-24+,39-26+,40-28+,41-30+,42-32+,43-34+

InChI key

ICFIZJQGJAJRSU-SGHXUWJISA-N

General description

Coenzyme Q (CoQ) is a natural compound, which is broadly distributed in living organisms. It is a constituent of the electron transfer system. CoQ contains a benzoquinone moiety and an isoprenoid side chain.[1]

Application

CoQ8 has been used as a ligand to activate peroxisome proliferator-activated receptor (PPAR).[2]

Packaging

2 mL Amber Glass Sealed Ampule (900151P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of coenzymeQ10 by recombinant E. coli harboring the decaprenyl diphosphate synthase gene from Sinorhizobium meliloti
Seo MJ, et al.
Journal of microbiology and biotechnology, 16(6), 933-938 (2006)
Idebenone and coenzyme Q10 are novel PPAR alpha gamma ligands, with potential for treatment of fatty liver diseases
Tiefenbach J, et al.
Disease models & mechanisms, 11(9), dmm034801-dmm034801 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service